Assay Details
Amplicon Length: | 98 |
Chromosome Location: | Chr. 19 - 53721372 - 53721372 [-] on 53721372 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-517b-3p | MIMAT0002857 |
| Human | hsa-miR-517-5p | MIMAT0002851 |
Stem-loop Details
Human

Genomic Map