Assay Details
Gene Family ID: | MIPF0000088, mir-224 |
Mature miRNA Sequence: | UAAGUCACUAGUGGUUCCGUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-224-5p | MIMAT0000671 |
Stem-loop Details
Mouse
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-224-5p |
|---|---|
| miRBase Accession Number: | MI0000711 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAAGUCACUAGUGGUUCCGUU |
| Species: | Mouse |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000088, mir-224 |
Mature miRNA Sequence: | UAAGUCACUAGUGGUUCCGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-224-5p | MIMAT0000671 |