Assay Details
Gene Family ID: | MIPF0000097, mir-338 |
Mature miRNA Sequence: | AACAAUAUCCUGGUGCUGAGUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-338-5p | MIMAT0004701 |
| Mouse | mmu-miR-338-5p | MIMAT0004647 |
| Rat | rno-miR-338-5p | MIMAT0004646 |
| Anolis carolinensis | aca-miR-338-5p | MIMAT0021937 |
| Columba livia | cli-miR-338b-5p | MIMAT0038620 |
| Horse | eca-miR-338-5p | MIMAT0013035 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-338-5p | MIMAT0015840 |
| Rhesus monkey | mml-miR-338-5p | MIMAT0006277 |
Stem-loop Details
Human
Mouse
Rat
Anolis carolinensis
Columba livia
Horse
Pongo pygmaeus
Rhesus monkey

Genomic Map