Assay Details
Amplicon Length: | 76 |
Chromosome Location: | Chr. 10 - 109195608 - 109195608 [+] on 109195608 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-3065-5p | MIMAT0017839 |
Rat | rno-miR-3065-3p | MIMAT0017840 |
Rat | rno-miR-338-3p | MIMAT0000581 |
Rat | rno-miR-338-5p | MIMAT0004646 |
Stem-loop Details
Rat
