Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | GAAGGCGCUUCCCUUUAGAGCG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-525-3p | MIMAT0002839 |
| Gorilla gorilla | ggo-miR-525-3p | MIMAT0036413 |
| Pan troglodytes | ptr-miR-525 | MIMAT0008209 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes

Genomic Map