Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAAGUAAUCGCGGUUUUUGUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548h-5p | MIMAT0005928 |
Callithrix jacchus | cja-miR-548f | MIMAT0039576 |
Pan troglodytes | ptr-miR-548h | MIMAT0008222 |
Stem-loop Details
Human
Callithrix jacchus
Pan troglodytes
