Assay Details
Gene Family ID: | MIPF0000558, mir-1307 |
Mature miRNA Sequence: | ACUCGGCGUGGCGUCGGUCGUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1307-3p | MIMAT0005951 |
| Bovine | bta-miR-1307 | MIMAT0009969 |
| Dog | cfa-miR-1307 | MIMAT0006665 |
| Pan troglodytes | ptr-miR-1307 | MIMAT0008024 |
| Pig | ssc-miR-1307 | MIMAT0013936 |
| Pongo pygmaeus | ppy-miR-1307 | MIMAT0016227 |
Stem-loop Details
Human
Bovine
Dog
Pan troglodytes
Pig
Pongo pygmaeus

Genomic Map