Assay Details
Gene Family ID: | MIPF0000531, mir-1306 |
Mature miRNA Sequence: | ACGUUGGCUCUGGUGGUGAUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1306-3p | MIMAT0009411 |
| Columba livia | cli-miR-1306-3p | MIMAT0038671 |
| Cricetulus griseus | cgr-miR-1306-3p | MIMAT0023753 |
| Gorilla gorilla | ggo-miR-1306 | MIMAT0024225 |
| Pig | ssc-miR-1306-3p | MIMAT0013938 |
| Pteropus alecto | pal-miR-1306-3p | MIMAT0040382 |
| View More Rows | ||
| Tupaia chinensis | tch-miR-1306-3p | MIMAT0036497 |
Stem-loop Details
Mouse
Columba livia
Cricetulus griseus
Gorilla gorilla
Pig
Pteropus alecto
Tupaia chinensis

Genomic Map