Assay Details
Gene Family ID: | MIPF0000199, mir-331 |
Mature miRNA Sequence: | GGUCUUGUUUGGGUUUGUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-331-5p | MIMAT0017033 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-331-5p |
|---|---|
| miRBase Accession Number: | MI0000608 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGUCUUGUUUGGGUUUGUU |
| Species: | Rat |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000199, mir-331 |
Mature miRNA Sequence: | GGUCUUGUUUGGGUUUGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-331-5p | MIMAT0017033 |