Assay Details
Amplicon Length: | 106 |
Chromosome Location: | Chr. 7 - 34881443 - 34881443 [+] on 34881443 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-331-3p | MIMAT0000570 |
| Rat | rno-miR-331-5p | MIMAT0017033 |
Stem-loop Details
Rat

Genomic Map