Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | CGGUUGACCAUGGUGUGUACG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1197-5p | MIMAT0017331 |
| Dasypus novemcinctus | dno-miR-1197-5p | MIMAT0048029 |
| Guinea pig | cpo-miR-1197-5p | MIMAT0047403 |
| Rabbit | ocu-miR-1197-5p | MIMAT0048583 |
Stem-loop Details
Mouse
Dasypus novemcinctus
Guinea pig
Rabbit

Genomic Map