Assay Details
Amplicon Length: | 67 |
Chromosome Location: | Chr. 12 - 109712112 - 109712112 [+] on 109712112 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1197-3p | MIMAT0005858 |
| Mouse | mmu-miR-1197-5p | MIMAT0017331 |
| Mouse | mmu-miR-323-3p | MIMAT0000551 |
| Mouse | mmu-miR-323-5p | MIMAT0004638 |
Stem-loop Details
Mouse

Genomic Map