Assay Details
Gene Family ID: | MIPF0000288, mir-374 |
Mature miRNA Sequence: | CUUAGCACGUUGUAUUAUUAUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-374-3p | MIMAT0017223 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-374-3p |
|---|---|
| miRBase Accession Number: | MI0003552 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUUAGCACGUUGUAUUAUUAUU |
| Species: | Rat |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000288, mir-374 |
Mature miRNA Sequence: | CUUAGCACGUUGUAUUAUUAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-374-3p | MIMAT0017223 |