Assay Details
Amplicon Length: | 73 |
Chromosome Location: | Chr. X - 74444000 - 74444000 [-] on 74444000 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-374-5p | MIMAT0003208 |
| Rat | rno-miR-374-3p | MIMAT0017223 |
Stem-loop Details
Rat

Genomic Map