Assay Details
Gene Family ID: | MIPF0000031, mir-196 |
Mature miRNA Sequence: | UCGACAGCACGACACUGCCUUCA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-196b-3p | MIMAT0017171 |
Dasypus novemcinctus | dno-miR-196b-3p | MIMAT0047732 |
Guinea pig | cpo-miR-196b-3p | MIMAT0047083 |
Rabbit | ocu-miR-196b-3p | MIMAT0048316 |
Stem-loop Details
Rat
Dasypus novemcinctus
Guinea pig
Rabbit
