Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. 4 - 82284501 - 82284501 [-] on 82284501 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-196b-5p | MIMAT0001082 |
Rat | rno-miR-196b-3p | MIMAT0017171 |
Stem-loop Details
Rat
