Assay Details
Amplicon Length: | 91 |
Chromosome Location: | Chr. 2 - 189421583 - 189421583 [+] on 189421583 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-190b-3p | MIMAT0017298 |
Rat | rno-miR-190b-5p | MIMAT0005302 |
Stem-loop Details
Rat
