Assay Details
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | UGAUAUGUUUGAUAUUAGGUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-190b-5p | MIMAT0005302 |
Xenopus tropicalis | xtr-miR-190 | MIMAT0025392 |
Stem-loop Details
Rat
Xenopus tropicalis
