Assay Details
Amplicon Length: | 139 |
Chromosome Location: | Chr. 9 - 26795504 - 26795504 [-] on 26795504 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-133b-5p | MIMAT0017205 |
| Rat | rno-miR-133b-3p | MIMAT0003126 |
Stem-loop Details
Rat

Genomic Map