Assay Details
Amplicon Length: | 99 |
Chromosome Location: | Chr. X - 158585413 - 158585413 [-] on 158585413 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-351-3p | MIMAT0017041 |
| Rat | rno-miR-351-5p | MIMAT0000608 |
Stem-loop Details
Rat

Genomic Map