Assay Details
Gene Family ID: | MIPF0000244, mir-351 |
Mature miRNA Sequence: | GGUCAAGAGGCGCCUGGGAAC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-351-3p | MIMAT0017042 |
| Rat | rno-miR-351-3p | MIMAT0017041 |
Stem-loop Details
Mouse
Rat
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-351* (v17)
Rat: rno-miR-351* (v18)
Rat: rno-miR-351* (v18)

Genomic Map