Assay Details
Amplicon Length: | 112 |
Chromosome Location: | Chr. X - 21583028 - 21583028 [-] on 21583028 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-let-7f-5p | MIMAT0000778 |
Rat | rno-let-7f-2-3p | MIMAT0017090 |
Stem-loop Details
Rat
