Assay Details
Amplicon Length: | 109 |
Chromosome Location: | Chr. X - 158585078 - 158585078 [-] on 158585078 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-503-5p | MIMAT0003213 |
| Rat | rno-miR-503-3p | MIMAT0017224 |
Stem-loop Details
Rat

Genomic Map