Assay Details
Amplicon Length: | 86 |
Chromosome Location: | Chr. X - 151272554 - 151272554 [+] on 151272554 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3551-5p | MIMAT0017809 |
| Rat | rno-miR-3551-3p | MIMAT0017810 |
| Rat | rno-miR-471-3p | MIMAT0017310 |
| Rat | rno-miR-471-5p | MIMAT0005318 |
Stem-loop Details
Rat

Genomic Map