Assay Details
Amplicon Length: | 81 |
Chromosome Location: | Chr. X - 114002770 - 114002770 [-] on 114002770 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-652-3p | MIMAT0005342 |
| Rat | rno-miR-652-5p | MIMAT0017321 |
Stem-loop Details
Rat

Genomic Map