Assay Details
Gene Family ID: | MIPF0000333, mir-652 |
Mature miRNA Sequence: | AAUGGCGCCACUAGGGUUGUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-652-3p | MIMAT0003322 |
| Mouse | mmu-miR-652-3p | MIMAT0003711 |
| Rat | rno-miR-652-3p | MIMAT0005342 |
| Bovine | bta-miR-652 | MIMAT0024578 |
| Callithrix jacchus | cja-miR-652a | MIMAT0039358 |
| Cricetulus griseus | cgr-miR-652-3p | MIMAT0023992 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-652-3p | MIMAT0047987 |
| Gorilla gorilla | ggo-miR-652 | MIMAT0024153 |
| Horse | eca-miR-652 | MIMAT0013240 |
| Pan troglodytes | ptr-miR-652 | MIMAT0008302 |
| Pongo pygmaeus | ppy-miR-652 | MIMAT0016085 |
| Rhesus monkey | mml-miR-652 | MIMAT0006493 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Cricetulus griseus
Dasypus novemcinctus
Gorilla gorilla
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-652 (v17)
Mouse: mmu-miR-652 (v17)
Rat: rno-miR-652 (v18)
Mouse: mmu-miR-652 (v17)
Rat: rno-miR-652 (v18)

Genomic Map