Assay Details
Amplicon Length: | 65 |
Chromosome Location: | Chr. 13 - 103192831 - 103192831 [-] on 103192831 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-664-2-5p | MIMAT0017229 |
Rat | rno-miR-664-3p | MIMAT0003382 |
Stem-loop Details
Rat
