Assay Details
Amplicon Length: | 73 |
Chromosome Location: | Chr. 18 - 47487697 - 47487697 [-] on 47487697 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-664-1-5p | MIMAT0017228 |
Rat | rno-miR-664-3p | MIMAT0003382 |
Stem-loop Details
Rat
