Assay Details
Gene Family ID: | MIPF0000598, mir-1298 |
Mature miRNA Sequence: | CAUCUGGGCAACUGAUUGAACU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1298-3p | MIMAT0014810 |
| Guinea pig | cpo-miR-1298-3p | MIMAT0047418 |
| Rabbit | ocu-miR-1298-3p | MIMAT0048596 |
Stem-loop Details
Mouse
Guinea pig
Rabbit

Genomic Map