Assay Details
Amplicon Length: | 126 |
Chromosome Location: | Chr. X - 147064755 - 147064755 [-] on 147064755 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1298-5p | MIMAT0014809 |
| Mouse | mmu-miR-1298-3p | MIMAT0014810 |
Stem-loop Details
Mouse

Genomic Map