Assay Details
Gene Family ID: | MIPF0000316, mir-467 |
Mature miRNA Sequence: | UGUCUUGUGUGUGCAUGUUCAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-669e-5p | MIMAT0005853 |
Stem-loop Details
Mouse
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-669e-5p |
|---|---|
| Stem-loop Accession Number: | MI0006300 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUCUUGUGUGUGCAUGUUCAU |
| Species: | Mouse |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000316, mir-467 |
Mature miRNA Sequence: | UGUCUUGUGUGUGCAUGUUCAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-669e-5p | MIMAT0005853 |