Assay Details
Gene Family ID: | MIPF0001105, mir-3065 |
Mature miRNA Sequence: | UCAGCACCAGGAUAUUGUUGGGGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-3065-3p | MIMAT0017840 |
Stem-loop Details
Rat

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | rno-miR-3065-3p |
---|---|
Stem-loop Accession Number: | MI0015429 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UCAGCACCAGGAUAUUGUUGGGGA |
Species: | Rat |
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0001105, mir-3065 |
Mature miRNA Sequence: | UCAGCACCAGGAUAUUGUUGGGGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-3065-3p | MIMAT0017840 |