Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (±)-Abscisic acid | 14375-45-2 | 10 MM 1 ML
SDP

Supplier:  Medchemexpress LLC HYN254910MM/1ML

Encompass_Preferred

(±)-Abscisic acid is an orally active plant hormone that is also present in animals. It contributes to the regulation of glycemia in mammals.

  • Orally active plant hormone
  • Regulates glycemia in mammals
  • High purity: 99.88%
  • Molecular weight: 264.32
  • Chemical formula: C15H20O4
  • Appears as a white to off-white solid
  • For research use only

Catalog No. 50-004-20825


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.