Learn More
Medchemexpress LLC Beta-N-methylamino-L-alanine hydrochloride | 16012-55-8 | 98.0% | 154.60 | 25 MG
Supplier: Medchemexpress LLC 25MGHYW015546
Beta-N-methylamino-L-alanine hydrochloride is a neurotoxin produced by cyanobacteria. It activates mGluR3 and inhibits PKC, and can be used in the research of neurodegenerative diseases and immune diseases.
- Activates mGluR3
- Inhibits PKC
- Useful for research into neurodegenerative diseases
- Useful for research into immune diseases
- Inhibits melatonin synthesis
- Shows toxicity in PC12 cells
- Suppresses cell cycle progression at the G1/S checkpoint
- Perturbs alanine, aspartate, and glutamate metabolism pathways
- Induces developmental neurotoxicity
- Causes neurological and pathological phenotypes mimicking amyotrophic lateral sclerosis (ALS)
- Induces in vivo retinal cell death
- Produces oxidative damage in liver and kidney of rats