Learn More
ACROBiosystems Streptavidin Coated Plates, Clear, 12x8-Well Strips, White Frame (For Serological Testing) 1 plate
Supplier: ACROBiosystems SP131PLATE
The streptavidin coated plate is developed for immunoassay and ELISA of serum samples. The plate could be separated to 12× 8-well strips and the frame is white. Streptavidin (SA) has an extraordinarily high affinity for biotin with a dissociation constant (Kd) on the order of 10−14 mol/L, the Biotinylated molecules can bind to the SA irreversibly. Streptavidin has an isoelectric point of 5 to 6, resulting in low nonspecific interactions. The Streptavidin Coated Plates we provide are easy to use and widely available for applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.