Learn More
Charles River Laboratories 0.03 Lysate 50-test vial (5.2 mL), 6 pack
Supplier:  Charles River Laboratories R15003PK/DEL
Encompass
Endosafe Rapid LAL Single-Test Vials contain gel-clot LAL to provide a simple, self-contained option for routine endotoxin detection in research samples, renal dialysis process water and dialysate solutions.
Catalog No. NC0375709