Learn More
BioSpec Products 0.1 mm Glass Beads 1 lb bottle
SDP
Supplier:  BioSpec Products 11079101
Encompass_Preferred
Glass has a density of 2.5 g/cc and is the most commonly used bead media for "beadbeating."  Beads are sold in one pound bottles with easy pour spouts.  Listed bead sizes are median (average) values.  Because beads are sorted by sieves, bead size varies between +/-10% of a listed value 1 lb bottle
Catalog No. NC0268065