Learn More
Boston Bioproducts Inc Tris Buffered Saline-Tween, TBST (10X, with 1% Tween-20, pH 7.4) - 1L
Supplier:  Boston Bioproducts Inc IBB1801L
Encompass_Preferred
10X Tris Buffered Saline - Tween (TBST) is a buffer-surfactant solution supporting a stable pH range between 7.4 and 8.0. When combined in a variable ratio, TBST is an effective wash buffer in Western blotting, in situ hybridization procedures, and immunohistochemistry staining, as it minimizes non specific binding of the materials without disrupting antigen-antibody binding. TBST is also used as an antibody diluent in immunoassays, and supports an array of molecular biology techniques.
Catalog No. NC9336835