Learn More
Teknova 0.1M Sodium Citrate, pH 3.6. 1000mL, Endotoxin Tested. Sterile. 2 pack.
SDP
Supplier:  Teknova S84402PK
Encompass
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.
Catalog No. 50-274-5819