Learn More
Chemglass Life Sciences Chemglass 0.1 mL 96-Well PCR Plate, Full Skirt, Clear, H1 Notch
SDP
Supplier:  Chemglass Life Sciences CGN3617F01
Encompass_Preferred
This 0.1 mL 96-well PCR plate from Chemglass is a clear, full-skirt polypropylene plate designed for low-volume PCR and reliable thermal cycling in automated workflows. It provides a standard 96-well format with an H1 orientation notch for consistent plate alignment.
  • 0.1 mL well volume.
  • 96 wells in standard footprint.
  • Full skirt for stability and automated handling.
  • H1 notch for plate orientation.
  • Polypropylene construction, RNase/DNase/pyrogen free.
  • Clear color suitable for optical assays.
Catalog No. 50-273-3416