Learn More
Chemglass Life Sciences Nest 0.1 mL 96-Well PCR Plate, No Skirt, Natural - CLS-3616-01N
SDP
Supplier:  Chemglass Life Sciences CLS361601N
Encompass_Preferred
This 0.1 mL, 96-well PCR plate features a non-skirted design and a natural (uncolored) finish, intended for low-volume PCR applications where a non-skirted format is required.
  • 0.1 mL well volume.
  • 96-well plate format.
  • No skirt (non-skirted) design.
  • Natural (uncolored) finish.
  • Typical material: polypropylene (natural, uncolored labware).
Catalog No. 50-273-3163