Learn More
Thomas Scientific GelRed Nucleic Acid Gel Stain, 10,000X in Water, 0.1mL
Supplier: Thomas Scientific C755G19
Phenix offers GelRed® 10,000X in water and GelRed® 10,000X in DMSO. GelRed® in water is a newer, safer formulation and recommended format. We continue to offer GelRed® in DMSO for established users who do not wish to alter their protocols. Replaces EtBr. Excites at 302/312nm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.