Learn More
Chemglass Life Sciences Chemglass 0.1 mL PCR 8-Strip Tubes, Clear, Nest
SDP
Supplier:  Chemglass Life Sciences CGN3612C01
Encompass_Preferred
Chemglass 0.1 mL PCR 8-Strip Tubes (nest) are low-profile, clear polypropylene tubes designed for PCR workflows. They minimize condensate and evaporation during thermal cycling, are free of DNase, RNase, human DNA, and PCR inhibitors, and withstand autoclaving.
  • 0.1 mL capacity per tube.
  • 8-strip configuration in a nest format.
  • Clear, low-profile design to minimize condensation and evaporation.
  • USP grade VI polypropylene construction.
  • Non-sterile; free of DNase, RNase, human DNA, and PCR inhibitors.
  • Autoclavable for convenient decontamination.
  • Intended for PCR applications.
Catalog No. 50-273-3423