Learn More
Chemglass Life Sciences Chemglass 0.1 mL PCR 8-Strip Tubes, White, 8-Strip (CGN-3612-W-01)
SDP
Supplier:  Chemglass Life Sciences CGN3612W01
Encompass_Preferred
0.1 mL PCR 8-strip tubes in white designed for reduced evaporation and compatibility with common thermal cyclers. Made from USP grade VI polypropylene, these low-profile tubes are free of DNase, RNase, human DNA, and common PCR inhibitors, and accept standard 8-strip caps.
  • 0.1 mL volume.
  • Eight-tube strip format for multi-sample PCR.
  • White color for easy sample identification.
  • USP grade VI polypropylene construction.
  • Compatible with domed or flat 8-strip caps (sterile or non-sterile).
  • Free of DNase, RNase, human DNA, and PCR inhibitors.
  • Autoclavable.
  • Low-profile design reduces condensate and evaporation.
  • Pack quantity code: C125.
Catalog No. 50-273-3421