Learn More
Chemglass Life Sciences Chemglass 0.1 mL PCR 8-Strip Tubes, White, Sterile (CGN-3612-W-02)
SDP
Supplier:  Chemglass Life Sciences CGN3612W02
Encompass_Preferred
Chemglass 0.1 mL PCR 8-Strip Tubes are sterile, white polypropylene tubes supplied in nested 8-tube strips for PCR and real-time thermal cycler applications. Thin-walled construction improves thermal transfer, and leak-proof caps reduce reagent loss during cycling.
  • 0.1 mL tube volume
  • 8 tubes per strip
  • Sterile, white polypropylene construction
  • Thin-walled design for improved thermal transfer
  • Leak-proof caps with less than 0.2% reaction loss
  • Compatible with most thermal cyclers and real-time PCR instruments
  • Supplied in nested 8-strip packaging
Catalog No. 50-273-3420