Learn More
Pacer Global LLC 0.2 ml 96-Well Skirted PCR Plate, Polypropylene, Non-sterile, Red, 25 per pack
SDP
Supplier:  Pacer Global LLC MLSP96FSR2/DEL
Encompass_Preferred
96-well skirted PCR Plate. Polypropylene. 200uL working Volume. Non-Sterile. Red Color. Certified RNase/DNase Free. Certified PCR Inhibitor Free. Certified Human DNA Free. Certified Pyrogen Free. Non-sterile. Thin wall design to maximize PCE efficiency. Compatible with most robotics. 25 plates/pack.
Catalog No. NC2065668