Learn More
Chemglass Life Sciences 0.2 mL 96-Well PCR Plate, No Skirt, Elevated Wells, Clear, H12 Notch, 25/Pack
SDP
Supplier:  Chemglass Life Sciences CGN3617E01
Encompass_Preferred
Chemglass 0.2 mL 96-well PCR plate with elevated wells and no skirt. Made from polypropylene and supplied 25 per pack, the plate is RNase-, DNase-, and pyrogen-free and is autoclavable; the cut corner at H12 provides orientation and the no-skirt format allows cutting to custom plate formats.
  • Polypropylene construction.
  • 0.2 mL well volume; 96 wells.
  • Elevated wells for improved sealing and pipetting.
  • No skirt; plate can be cut to any desired format.
  • Cut corner at H12 for orientation.
  • RNase-, DNase-, and pyrogen-free.
  • Autoclavable.
  • Clear; 25 plates per pack.
Catalog No. 50-273-3417