Learn More
Chemglass Life Sciences 0-4mm, 90°, Glass Only
SDP
Supplier:  Chemglass Life Sciences CGK50301
Encompass_Preferred
0-4mm, 90, Glass Only. Hi-vac valve with two sidearms sealed to the barrel at a 90 angle to one another. Designed with a precision external thread for fine control.
Catalog No. 50-153-2012