Learn More
Teknova 0.5M Sodium Citrate Buffer, pH 4.5. 250mL, RNase/DNase Tested, Sterile. 12 pack.
SDP
Supplier:  Teknova S851112PK
Encompass
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.
Catalog No. 50-238-7482