Learn More
Chemglass Life Sciences Chemglass 0.5 mL Cryogenic Vial, Self-Standing, Internal Thread, Sterile, Racked (1,600/case)
SDP
Supplier:  Chemglass Life Sciences CGN4766006
Encompass_Preferred
Self-standing 0.5 mL cryogenic vial with internal thread, supplied sterile and racked for efficient handling and cryogenic storage. Manufactured under cleanroom conditions and sterilized by E-beam for secure, long-term sample preservation.
  • 0.5 mL capacity.
  • Self-standing, round bottom design for stability.
  • Internal thread for secure closure.
  • Sterile; E-beam sterilized.
  • Made from USP 6 grade raw materials.
  • Autoclavable.
  • Packaging: 10 x 10 vials per rack, 16 racks per case (1,600 vials/case).
  • Non-pyrogenic; DNase/RNase free.
  • Recommended storage in the gas phase of liquid nitrogen.
  • Manufactured in a 100,000 grade clean room; ISO 9001 and ISO 13485 certified.
Catalog No. 50-273-3347